Läs mer
Ethyl Acid Tartrate, TRC
Produktkod. 30371460
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30371460 25 mg
1 options
Produktkod. 30371460 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE67962025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2R,3R)-4-Ethoxy-2,3-dihydroxy-4-oxobutanoic acid
CAS 608-89-9
Anteckningar om renhetsgrad HPLC
Molekylformel C6H10O6
Kvantitet 25 mg
Synonym Ethyl acid tartrate, Monoethyl tartrate
InChI formel InChI=1S/C6H10O6/c1-2-12-6(11)4(8)3(7)5(9)10/h3-4,7-8H,2H2,1H3,(H,9,10)/t3-,4-/m1/s1
LEDER OC([C@H](O)[C@@H](O)C(OCC)=O)=O
Molekylvikt (g/mol) 178.14
Formel vikt 178.0477