Läs mer
Ethyl-alpha-13C-benzene, CDN
Produktkod. 30309897
Change view
Kvantitet:
0.5 g
Förpackningsstorlek:
0.5g
Produktkod. Kvantitet unitSize
30309897 0.5 g 0.5g
1 options
Produktkod. 30309897 Leverantör CDN Leverantörsnummer CDNC31650.5G
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethylbenzene-alpha-13 C
CAS 287399-32-0
Molekylformel 13 C C7 H10
Kvantitet 0.5 g
Synonym Benzene, ethyl-1-13 C- (9 CI), Ethyl-1-13 C-benzene, (Ethyl-α-13 C)benzene, Ethylbenzene-13 C, Ethylbenzene-α-13 C, Ethyl-α-13 C-benzene
InChI formel InChI=1 S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i2+1
LEDER C[13 CH2]c1ccccc1
IUPAC-namn (113 C)ethylbenzene
Molekylvikt (g/mol) 107.1577
Formel vikt 107.0816 g/mol
Procent renhet 99 atom % 13 C, min 98% Chemical Purity