Läs mer
Ethyl alpha-(p-Methoxyphenyl)-beta-(dimethylamino)propionate, TRC
Produktkod. 30006531
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006531 50 mg
30004855 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006531 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92505550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 323176-93-8
Molekylformel C14 H21 N O3
Molekylvikt (g/mol) 251.32
Synonym Benzeneacetic acid, α-[(dimethylamino)methyl]-4-methoxy-, ethyl ester, Ethyl α-(p-methoxyphenyl)-β-(dimethylamino)propionate, α-[(Dimethylamino)methyl]-4-methoxybenzeneacetic Acid Ethyl Ester, Venlafaxine EP Impurity B;
IUPAC-namn ethyl 3-(dimethylamino)-2-(4-methoxyphenyl)propanoate
LEDER CCOC(=O)C(CN(C)C)c1ccc(OC)cc1
Specifikationer
Kemiskt namn eller material Ethyl alpha-(p-Methoxyphenyl)-beta-(dimethylamino)propionate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C14H21NO3/c1-5-18-14(16)13(10-15(2)3)11-6-8-12(17-4)9-7-11/h6-9,13H,5,10H2,1-4H3
Formel vikt 251.1521
Procent renhet >95%