Learn More
Ethyl b-D-Thiogalactopyranoside, TRC
Product Code. 30376381
Change view
Click to view available options
Kvantitet:
5 g
Product Code. Kvantitet
30376381 5 g
1 options
Product Code. 30376381 Supplier Toronto Research Chemicals Supplier No. TRCE9267755G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Ethyl beta-D-Thiogalactopyranoside
CAS 56245-60-4
Molekylformel C8 H16 O5 S
Kvantitet 5 g
InChI formel InChI=1S/C8H16O5S/c1-2-14-8-7(12)6(11)5(10)4(3-9)13-8/h4-12H,2-3H2,1H3/t4-,5+,6+,7-,8+/m1/s1
LEDER CCS[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
IUPAC-namn (2S,3R,4S,5R,6R)-2-ethylsulfanyl-6-(hydroxymethyl)oxane-3,4,5-triol
Molekylvikt (g/mol) 224.27
Formel vikt 224.0718