missing translation for 'onlineSavingsMsg'
Läs mer

Etyl Beta-tioglucopyranosid, TRC

Produktkod. 30373775
Change view
Click to view available options
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30373775 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373775 Supplier Toronto Research Chemicals Supplier No. TRCE926995500MG
 Begränsad artikel
Beräknad leverans: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Högrena organiska molekyler och analytiska standarder, strategiskt levererade världen över för att möjliggöra innovation och kommersiell framgång.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Ethyl beta-D-Thioglucopyranoside
CAS 7473-36-1
Molekylformel C8 H16 O5 S
Kvantitet 500 mg
InChI formel InChI=1S/C8H16O5S/c1-2-14-8-7(12)6(11)5(10)4(3-9)13-8/h4-12H,2-3H2,1H3/t4-,5-,6+,7-,8+/m1/s1
LEDER CCS[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
IUPAC-namn (2S,3R,4S,5S,6R)-2-etylsulfanyl-6-(hydroxymetyl)oxan-3,4,5-triol
Molekylvikt (g/mol) 224.27
Formel vikt 224.0718
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.