missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Ethyl Biscoumacetate, Mikromol™
Beskrivning
Ethyl Biscoumacetate
Specifikationer
Specifikationer
| Kemiskt namn eller material | Ethyl Biscoumacetate |
| CAS | 548-00-5 |
| Analyt- eller komponentnamn | Ethyl Biscoumacetate |
| Frakt skick | Room Temperature |
| Molekylformel | C22 H16 O8 |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Acetic acid, bis(4-hydroxy-2-oxo-2H-1-benzopyran-3-yl)-, ethyl ester (6CI,7CI,8CI), 3,3'-(Carboxymethylene)bis(4-hydroxycoumarin) ethyl ester, BOEA, Bis(4-hydroxy-2-oxo-2H-1-benzopyran-3-yl)acetic acid ethyl ester, Bis(4-hydroxy-3-coumarin)acetic acid ethyl ester, Dicumacyl, Ethyl 4,4'-dihydroxydicoumarinyl-3,3'-acetate, Ethyl bis(4-hydroxycoumarinyl)acetate, Ethyl biscoumacetate, NSC 36366, Neodicoumarin, Neodicoumarol, Neodicumarinum, Neodicumarol, Pelentan, Stabilene, Trombarin, Trombil, Trombolysan, Tromexan, Tromexan ethyl acetate, 2H-1-Benzopyran-3-acetic acid, 4-hydroxy-alpha-(4-hydroxy-2-oxo-2H-1-benzopyran-3-yl)-2-oxo-, ethyl ester |
| InChI formel | InChI=1S/C22H16O8/c1-2-28-20(25)15(16-18(23)11-7-3-5-9-13(11)29-21(16)26)17-19(24)12-8-4-6-10-14(12)30-22(17)27/h3-10,15,23-24H,2H2,1H3 |
| LEDER | CCOC(=O)C(C1=C(O)c2ccccc2OC1=O)C3=C(O)c4ccccc4OC3=O |
| IUPAC-namn | ethyl 2,2-bis(4-hydroxy-2-oxochromen-3-yl)acetate |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?