missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cinnamate, 98% trans, Thermo Scientific Chemicals

Catalog Number p-4510173
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
103-36-6
C11H12O2
176.21
KBEBGUQPQBELIU-CMDGGOBGSA-N
ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate
637758
CHEBI:4895
ethyl (E)-3-phenylprop-2-enoate
CCOC(=O)C=CC1=CC=CC=C1
This item is not returnable. View return policy
103-36-6
C11H12O2
176.21
KBEBGUQPQBELIU-CMDGGOBGSA-N
ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate
637758
CHEBI:4895
ethyl (E)-3-phenylprop-2-enoate
CCOC(=O)C=CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 1mg KOH/g max.
Melting Point 6.5°C to 7.5°C
Color Colorless to Yellow
Density 1.0460g/mL
Boiling Point 271.0°C
Flash Point 135°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH=CHCO2C2H5
Refractive Index 1.5580 to 1.5610
Quantity 100 g
Merck Index 15,2299
Specific Gravity 1.046
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 176.21
Percent Purity 98% trans
Physical Form Liquid
Chemical Name or Material Ethyl cinnamate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'