Läs mer
Ethyl Cyanoacetate-13C2, TRC
Produktkod. 30377398
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30377398 250 mg
1 options
Produktkod. 30377398 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE678462250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl Cyanoacetate-13C2
CAS 93664-96-1
Molekylformel C313C2H7NO2
Kvantitet 250 mg
Synonym 2-Cyano-acetic Acid Ethyl Ester-13C2, (Ethoxycarbonyl)acetonitrile-13C2, Cyanoacetic alphacid epsilonthyl epsilonster-13C2, Cyanoacetic Ester-13C2, Ethyl 2-Cyanoacetate-13C2, Ethyl Cyanacetate-13C2, Ethyl Cyanoacetate-13C2, Ethyl Cyanoethanoate-13C2, Ethyl alpha-Cyanoacetate-13C2, Malonic Acid Ethyl Ester Nitrile-13C2, NSC 8844-13C2
InChI formel InChI=1S/C5H7NO2/c1-2-8-5(7)3-4-6/h2-3H2,1H3/i4+1,5+1
LEDER CCO[13C](=O)C[13C]#N
IUPAC-namn ethyl 2-(azanylidyne(1^{13}C)methyl)acetate
Molekylvikt (g/mol) 115.1
Formel vikt 115.0544