Läs mer
Ethyl Cyanoacetate-3-13C, TRC
Produktkod. 30375017
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30375017 1 g
1 options
Produktkod. 30375017 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9055571G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl Cyanoacetate-3-13C
CAS 698387-41-6
Molekylformel C413CH7NO2
Kvantitet 1 g
Synonym 2-Cyano-acetic Acid Ethyl Ester-13C, (Ethoxycarbonyl)acetonitrile-13C, Cyanoacetic Acid Ethyl Ester-13C, Cyanoacetic Ester-13C, Ethyl 2-Cyanoacetate-13C, Malonic Acid Ethyl Ester Nitrile-13C, NSC 8844-13C
InChI formel InChI=1S/C5H7NO2/c1-2-8-5(7)3-4-6/h2-3H2,1H3/i4+1
LEDER N#[13C]CC(OCC)=O
IUPAC-namn ethyl 2-(cyano)acetate
Molekylvikt (g/mol) 114.11
Formel vikt 114.05