Läs mer
Ethyl Diazoacetate (>90%), TRC
Produktkod. 30378635
Change view
Kvantitet:
5 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378635 5 g
30372814 10 g
30372400 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30378635 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9156005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 623-73-4
Molekylformel C4H6N2O2
Molekylvikt (g/mol) 114.1
Synonym 2-Diazo-1-ethoxyethanone, DAAE, Diazoacetic Acid Ethyl Ester, Diazoacetic Ester, Ethoxycarbonyldiazomethane, Ethyl 2-Diazoacetate, Ethyl 2-Diazoethanoate, Ethyl Diazoacetate, NSC 79147, 2-Diazoacetic Acid Ethyl Ester
LEDER CCOC(=O)C=[N+]=[N-]
Specifikationer
Kemiskt namn eller material Ethyl Diazoacetate
Kvantitet 5 g
InChI formel InChI=1S/C4H6N2O2/c1-2-8-4(7)3-6-5/h3H,2H2,1H3
Formel vikt 114.0429