Läs mer
Ethyl (E)-3-(2-Pyridyl)acrylate, TRC
Produktkod. 30378509
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30378509 50 mg
1 options
Produktkod. 30378509 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92567050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl (E)-3-(2-Pyridyl)acrylate
CAS 70526-11-3
Molekylformel C10 H11 N O2
Kvantitet 50 mg
Synonym 2-Propenoic acid, 3-(2-pyridinyl)-, ethyl ester, (2E)-, 2-Propenoic acid, 3-(2-pyridinyl)-, ethyl ester, (E)-, (2E)-3-(2-Pyridinyl)-2-propenoic acid ethyl ester, Ethyl (E)-3-(pyridin-2-yl)-2-propenoate, Ethyl (E)-3-(pyridin-2-yl)acrylate, (2E)-3-(Pyridin-2-yl)acrylic acid ethyl ester, trans-3-(Pyridin-2-yl)acrylic acid ethyl ester
InChI formel InChI=1S/C10H11NO2/c1-2-13-10(12)7-6-9-5-3-4-8-11-9/h3-8H,2H2,1H3/b7-6+
LEDER CCOC(=O)\C=C\c1ccccn1
IUPAC-namn ethyl (E)-3-pyridin-2-ylprop-2-enoate
Molekylvikt (g/mol) 177.2
Formel vikt 177.079