Läs mer
Ethyl Glycodeoxycholate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Glycodeoxycholic Acid Ethyl Ester |
| CAS | 70779-06-5 |
| Molekylformel | C28 H47 N O5 |
| Kvantitet | 100 mg |
| Synonym | Glycine, N-[(3α,5β,12α)-3,12-dihydroxy-24-oxocholan-24-yl]-, ethyl ester, Ethyl glycodeoxycholate, Glycodeoxycholic Acid Ethyl Ester |
| InChI formel | InChI=1S/C28H47NO5/c1-5-34-26(33)16-29-25(32)11-6-17(2)21-9-10-22-20-8-7-18-14-19(30)12-13-27(18,3)23(20)15-24(31)28(21,22)4/h17-24,30-31H,5-16H2,1-4H3,(H,29,32)/t17-,18-,19-,20+,21-,22+,23+,24+,27+,28-/m1/s1 |
| LEDER | CCOC(=O)CNC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C |
| IUPAC-namn | ethyl 2-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate |
| Molekylvikt (g/mol) | 477.68 |
| Formel vikt | 477.3454 |