missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl-m-Aminobenzoate, methanesulfonate salt, MP Biomedicals™
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
Specifications
Color | White |
Density | Ethyl-m-Aminobenzoate is mainly used as an anesthetic in fish,sharks,frogs |
Assay Percent Range | 0.993 |
Quantity | 5 g |
Formula Weight | 261.3 |
Physical Form | Powder |
Chemical Name or Material | Ethyl-m-Aminobenzoate, Methanesulfonate Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'