Läs mer
Ethyl Malonamate, TRC
Produktkod. 30340211
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30340211 100 mg
1 options
Produktkod. 30340211 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB433778100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl Malonamate
CAS 7597-56-0
Molekylformel C5H9NO3
Kvantitet 100 mg
Synonym Ethyl 3-Amino-3-oxopropanoate, Ethoxycarbonylacetamide, Ethyl Aminocarbonylacetate, Ethyl Carbamoylacetate
InChI formel InChI=1S/C5H9NO3/c1-2-9-5(8)3-4(6)7/h2-3H2,1H3,(H2,6,7)
LEDER CCOC(=O)CC(=O)N
IUPAC-namn ethyl 3-amino-3-oxopropanoate
Molekylvikt (g/mol) 131.13
Formel vikt 131.0582