Läs mer
Ethyl(methoxy)amine Hydrochloride, TRC
Produktkod. 30334758
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30334758 100 mg
1 options
Produktkod. 30334758 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB436928100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methoxyethanamine hydrochloride
CAS 1195657-97-6
Molekylformel C3 H9 N O . H Cl
Kvantitet 100 mg
Synonym Ethyl(methoxy)amine Hydrochloride
InChI formel InChI=1S/C3H9NO.ClH/c1-3-4-5-2;/h4H,3H2,1-2H3;1H
LEDER Cl.CCNOC
IUPAC-namn N-methoxyethanamine;hydrochloride
Molekylvikt (g/mol) 111.57
Formel vikt 111.0451