Learn More
Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 97%, tech., Thermo Scientific Chemicals
Catalog Number p-3736437
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
52763-21-0
C15H20ClNO3
297.78
MFCD00012792
UQOMEAWPKSISII-UHFFFAOYNA-N
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, ethyl n-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride, ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 1-benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hcl, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hydrochloride, 1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride, ethyl n-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, zlchem 420, pubchem19671
2723880
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride
[H+].[Cl-].CCOC(=O)C1CCN(CC2=CC=CC=C2)CC1=O
52763-21-0
C15H20ClNO3
297.78
MFCD00012792
UQOMEAWPKSISII-UHFFFAOYNA-N
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, ethyl n-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride, ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 1-benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hcl, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hydrochloride, 1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride, ethyl n-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, zlchem 420, pubchem19671
2723880
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride
[H+].[Cl-].CCOC(=O)C1CCN(CC2=CC=CC=C2)CC1=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 160°C to 165°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 297.77
Percent Purity 97%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl N-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.