Läs mer
Ethyl Nicotinate-1,2’,3’,4’,5’,6’-13C6 Hydrochloride Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Ethyl Nicotinate-1,2',3',4',5',6'-13C6 Hydrochloride Salt |
| CAS | 1346604-87-2 |
| Molekylformel | C213C6H10ClNO2 |
| Kvantitet | 0.5 mg |
| Synonym | 3-Pyridinecarboxylic Acid-13C6 Ethyl Ester Hydrochloride, Nicotinic Acid-13C6 Hydrochloride Ethyl Ester Hydrochloride, 3-(Ethoxycarbonyl)pyridine-13C6 Hydrochloride, 3-Carbethoxypyridine-13C6 Hydrochloride, Ba 2673-13C6 Hydrochloride, Ethyl 3-Pyridinecarboxylate-13C6 Hydrochloride, Ignicut, Mucotherm-13C6, NSC 8872-13C6, Nicaethan-13C6, Nikethan-13C6, Nikithan-13C6, beta-Pyridinecarboxylic-13C6 Acid Ethyl Ester Hydrochloride |
| InChI formel | InChI=1S/C8H9NO2.ClH/c1-2-11-8(10)7-4-3-5-9-6-7;/h3-6H,2H2,1H3;1H/i3+1,4+1,5+1,6+1,7+1,8+1; |
| LEDER | Cl.CCO[13C](=O)[13c]1[13cH][13cH][13cH]n[13cH]1 |
| Molekylvikt (g/mol) | 193.58 |
| Formel vikt | 193.0601 |