missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oleate, 98%, mixture of homologeous fatty acid esters, Thermo Scientific Chemicals

Catalog Number p-4509554
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
111-62-6
C20H38O2
310.51
LVGKNOAMLMIIKO-QXMHVHEDSA-N
ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
5363269
CHEBI:84940
ethyl (Z)-octadec-9-enoate
CCCCCCCCC=CCCCCCCCC(=O)OCC
This item is not returnable. View return policy
111-62-6
C20H38O2
310.51
LVGKNOAMLMIIKO-QXMHVHEDSA-N
ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
5363269
CHEBI:84940
ethyl (Z)-octadec-9-enoate
CCCCCCCCC=CCCCCCCCC(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 0.5mg KOH/g max.
Density 0.8600g/mL
Boiling Point 190.0°C to 210.0°C (7.0 mmHg)
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 60% min. (oleic acid) (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7CHCH(CH2)7COOC2H5
Refractive Index 1.4490 to 1.4530
Quantity 250 g
Saponification Value 177 to 188mg KOH/g
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 310.51
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material Ethyl oleate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'