Läs mer
Ethyl p-Aminobenzoate-N-D-mannose, TRC
Produktkod. 30375271
Change view
Kvantitet:
20 mg
Produktkod. Kvantitet
30375271 20 mg
1 options
Produktkod. 30375271 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE67829520MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl p-Aminobenzoate-N-D-mannose
CAS 78216-92-9
Molekylformel C15 H21 N O7
Kvantitet 20 mg
Synonym 4-(D-Mannopyranosylamino)benzoic Acid Ethyl Ester, Ethyl p-Aminobenzoate-N-D-mannose
InChI formel InChI=1S/C15H21NO7/c1-2-22-15(21)8-3-5-9(6-4-8)16-14-13(20)12(19)11(18)10(7-17)23-14/h3-6,10-14,16-20H,2,7H2,1H3/t10-,11-,12+,13+,14?/m1/s1
LEDER CCOC(=O)c1ccc(NC2O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]2O)cc1
IUPAC-namn ethyl 4-[[(3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]amino]benzoate
Molekylvikt (g/mol) 327.33
Formel vikt 327.1318