Läs mer
Ethyl Paraben-13C6, TRC
Produktkod. 30008746
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008746 10 mg
30004983 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008746 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92547910MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2075718-25-9
Molekylformel C313C6H10O3
Molekylvikt (g/mol) 172.13
Synonym 4-hydroxybenzoic Acid Ethyl Ester-13C6, 4-(Ethoxycarbonyl)phenol-13C6, 4-Carbethoxyphenol-13C6, 4-Hydroxybenzoic Acid Ethyl Ester-13C6, Aseptin A-13C6, Aseptoform E-13C6, Easeptol-13C6, Ethyl 4-Hydroxybenzoate-13C6, Ethyl Butex-13C6, Mekkings E-13C6, Mycocten-13C6, NSC 23514-13C6, p-Carbethoxyphenol-13C6;
LEDER [H][13C]1=[13C](O)[13C]([H])=[13C]([H])[13C](C(OCC)=O)=[13C]1[H]
Specifikationer
Kemiskt namn eller material Ethyl Paraben-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3/i3+1,4+1,5+1,6+1,7+1,8+1
Procent renhet >95%