Läs mer
Ethyl (Pentafluorobenzoyl)acetate, TRC
Produktkod. 30369892
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30369892 50 mg
30374972 500 mg
30370846 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30369892 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE81801850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3516-87-8
Molekylformel C11H7O3F5
Molekylvikt (g/mol) 282.16
LEDER CCOC(=O)CC(=O)c1c(F)c(F)c(F)c(F)c1F
Specifikationer
Kemiskt namn eller material Ethyl (Pentafluorobenzoyl)acetate
Kvantitet 50 mg
InChI formel InChI=1S/C11H7F5O3/c1-2-19-5(18)3-4(17)6-7(12)9(14)11(16)10(15)8(6)13/h2-3H2,1H3
Formel vikt 282.0315