missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenoxyacetate, 98%, Thermo Scientific™

Catalog Number p-131886
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2555-49-9
C10H12O3
180.2
MFCD00026895
MGZFVSUXQXCEHM-UHFFFAOYSA-N
ethyl phenoxyacetate, acetic acid, phenoxy-, ethyl ester, phenoxyacetic acid ethyl ester, acetic acid, 2-phenoxy-, ethyl ester, phenoxy-acetic acid ethyl ester, phenoxyacetic acid, ethyl ester, ethyl2-phenoxyacetate, pubchem12485, acmc-1ckja, ethylphenoxyacetate
17365
ethyl 2-phenoxyacetate
CCOC(=O)COC1=CC=CC=C1
This item is not returnable. View return policy
2555-49-9
C10H12O3
180.2
MFCD00026895
MGZFVSUXQXCEHM-UHFFFAOYSA-N
ethyl phenoxyacetate, acetic acid, phenoxy-, ethyl ester, phenoxyacetic acid ethyl ester, acetic acid, 2-phenoxy-, ethyl ester, phenoxy-acetic acid ethyl ester, phenoxyacetic acid, ethyl ester, ethyl2-phenoxyacetate, pubchem12485, acmc-1ckja, ethylphenoxyacetate
17365
ethyl 2-phenoxyacetate
CCOC(=O)COC1=CC=CC=C1
Color Yellow
Density 1.1000g/mL
Boiling Point 136°C (19.0 mmHg)
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5035
Quantity 5 g
Specific Gravity 1.1
Solubility Information Solubility in water: immiscible in water
Formula Weight 180.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl phenoxyacetate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'