missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Phthalyl Ethyl Glycolate, TRC

Product Code. 30133615
Change view
Click to view available options
Kvantitet:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30133615 250 mg
30135317 2.5 g
17991175 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133615 Supplier Toronto Research Chemicals Supplier No. TRCE937355250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-72-0
Molekylformel C14H16O6
Molekylvikt (g/mol) 280.27
Synonym Phthalic Acid, Ethyl Ester-(ethyl glycolate), 1,2-Benzenedicarboxylic acid, 1-(2-ethoxy-2-oxoethyl) 2-ethyl ester (ACI), 1,2-Benzenedicarboxylic acid, 2-ethoxy-2-oxoethyl ethyl ester (9CI), Glycolic acid, ethyl ester, ethyl phthalate (8CI), Phthalic acid, ethyl ester, ester with ethyl glycolate (6CI, 7CI, 8CI), 2-O-(2-Ethoxy-2-oxoethyl) 1-O-ethyl benzene-1,2-dicarboxylate, Carbethoxymethyl ethyl phthalate, Diethyl o-carboxybenzoyloxyacetate, Ethoxycarbonylmethyl ethyl phthalate, Ethyl 2-ethoxy-2-oxoethyl phthalate, Ethyl carbethoxymethyl phthalate, Ethyl ethoxycarbonylmethyl phthalate, Ethyl phthalyl ethyl glycolate, Santicizer E-15, Phthalic Acid, Ethyl Ester-(ethyl glycolate), 1,2-Benzenedicarboxylic acid, 1-(2-ethoxy-2-oxoethyl) 2-ethyl ester (ACI), 1,2-Benzenedicarboxylic acid, 2-ethoxy-2-oxoethyl ethyl ester (9CI), Glycolic acid, ethyl ester, ethyl phthalate (8CI), Phthalic acid, ethyl ester, ester with ethyl glycolate (6CI, 7CI, 8CI), 2-O-(2-Ethoxy-2-oxoethyl) 1-O-ethyl benzene-1,2-dicarboxylate, Carbethoxymethyl ethyl phthalate, Diethyl o-carboxybenzoyloxyacetate, Ethoxycarbonylmethyl ethyl phthalate, Ethyl 2-ethoxy-2-oxoethyl phthalate, Ethyl carbethoxymethyl phthalate, Ethyl ethoxycarbonylmethyl phthalate, Santicizer E-15
IUPAC-namn 2-O-(2-ethoxy-2-oxoethyl) 1-O-ethyl benzene-1,2-dicarboxylate
LEDER CCOC(=O)COC(=O)c1ccccc1C(=O)OCC

Specifications

Kemiskt namn eller material Ethyl Phthalyl Ethyl Glycolate
Kvantitet 250 mg
InChI formel InChI=1S/C14H16O6/c1-3-18-12(15)9-20-14(17)11-8-6-5-7-10(11)13(16)19-4-2/h5-8H,3-4,9H2,1-2H3
Formel vikt 280.0947
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.