missing translation for 'onlineSavingsMsg'
Learn More

Ethyl piperidine-1-acetate, 98%, Thermo Scientific™

Catalog Number p-7021358
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
23853-10-3
C9H17NO2
171.24
MFCD00134496
LXZHLNWWDDNIOJ-UHFFFAOYSA-N
643466
ethyl 2-piperidin-1-ylacetate
CCOC(=O)CN1CCCCC1
This item is not returnable. View return policy
23853-10-3
C9H17NO2
171.24
MFCD00134496
LXZHLNWWDDNIOJ-UHFFFAOYSA-N
643466
ethyl 2-piperidin-1-ylacetate
CCOC(=O)CN1CCCCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.984
Boiling Point 109°C to 111°C (25 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.454
Quantity 5 g
Beilstein 123142
Synonym ethyl 1-piperidineacetate, ethyl piperidin-1-ylacetate, ethyl 2-piperidin-1-yl acetate, ethyl piperidine-1-acetate, 1-piperidineacetic acid, ethyl ester, ethyl 2-piperidylacetate, ethyl1-piperidineacetate, acmc-20al1z, ethyl 1-piperidine acetate, ethyl 2-piperidinylacetate
Formula Weight 171.24
Percent Purity 98%
Chemical Name or Material Ethyl piperidine-1-acetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'