Läs mer
Ethyl Propionylacetate-d3, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Ethyl Propionylacetate-d3 |
| CAS | 1346598-17-1 |
| Molekylformel | C7H9D3O3 |
| Kvantitet | 10 mg |
| Synonym | 3-Oxopentanoic Acid Ethyl Ester, 3-Oxovaleric Acid Ethyl Ester, 3-Oxopentanoic Acid Ethyl Ester, Ethyl 3-Ketopentanoate, Ethyl 3-Oxopentanoate, Ethyl 3-Oxovalerate, Ethyl Propanoylacetate, Ethyl Propioacetate, Ethyl .beta.-oxovalerate, NSC 7251, NSC 78662, Propionylacetic Acid Ethyl Ester |
| InChI formel | InChI=1S/C7H12O3/c1-3-6(8)5-7(9)10-4-2/h3-5H2,1-2H3/i1D3 |
| LEDER | [2H]C([2H])([2H])CC(=O)CC(=O)OCC |
| IUPAC-namn | ethyl 5,5,5-trideuterio-3-oxopentanoate |
| Molekylvikt (g/mol) | 147.19 |
| Formel vikt | 147.0975 |