Läs mer
Ethyl (S)-2-(Benzyloxy)propionate, TRC
Produktkod. 30375019
Change view
Kvantitet:
2.5 g
Förpackningsstorlek:
2.5g
Produktkod. Kvantitet unitSize
30375019 2.5 g 2.5g
1 options
Produktkod. 30375019 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9000102.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl (S)-2-(Benzyloxy)propionate
CAS 54783-72-1
Molekylformel C12 H16 O3
Kvantitet 2.5 g
Synonym Propanoic acid, 2-(phenylmethoxy)-, ethyl ester, (2S)-, Propanoic acid, 2-(phenylmethoxy)-, ethyl ester, (S)-, Ethyl (2S)-2-(phenylmethoxy)propanoate, (-)-(S)-Ethyl 2-(benzyloxy)propanoate, (-)-(S)-Ethyl 2-(phenylmethoxy)propanoate, (S)-2-Benzyloxypropionic acid ethyl ester, (S)-Ethyl 2-(benzyloxy)propanoate, Ethyl (S)-2-benzyloxypropionate, Ethyl (S)-2-(benzyloxy)propionate, Ethyl (2S)-2-(benzyloxy)propanoate
InChI formel InChI=1S/C12H16O3/c1-3-14-12(13)10(2)15-9-11-7-5-4-6-8-11/h4-8,10H,3,9H2,1-2H3/t10-/m0/s1
LEDER CCOC(=O)[C@H](C)OCc1ccccc1
IUPAC-namn ethyl (2S)-2-phenylmethoxypropanoate
Molekylvikt (g/mol) 208.25
Formel vikt 208.1099