Läs mer
Ethyl Thiamine Bromide Hydrobromide, TRC
Produktkod. 30134328
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134328 250 mg
30008423 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134328 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE926690250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6309-04-2
Molekylformel C13H19BrN4OS •HBr
Molekylvikt (g/mol) 359.298091
Synonym 3-[(4-Amino-2-ethyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methylthiazolium Bromide Hydrobromide, 2'-Ethylthiamine Bromide Hydrobromide, Ethylthiamin Bromide Hydrobromide, Ethylthiamine
LEDER OCCC1=C(C)[N+](CC2=C(N)N=C(CC)N=C2)=CS1.[Br].Br
Specifikationer
Kemiskt namn eller material Ethyl Thiamine Bromide Hydrobromide
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C13H19N4OS.2BrH/c1-3-12-15-6-10(13(14)16-12)7-17-8-19-11(4-5-18)9(17)2;;/h6,8,18H,3-5,7H2,1-2H3,(H2,14,15,16);2*1H/q+1;;/p-1
Formel vikt 437.9725
Procent renhet >95%