Learn More
Ethyl Thiophene-2-carboxylate, Mikromol™
Product Code. 30050756
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30050756 100 mg 100mg
1 options
Product Code. 30050756 Supplier Mikromol Supplier No. MM1081.02
 Restricted Item
Beräknad leverans: 27-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl Thiophene-2-carboxylate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl Thiophene-2-carboxylate
CAS 2810-04-0
Analyte or Component Names Ethyl Thiophene-2-carboxylate
Shipping Condition Room Temperature
Molecular Formula C7 H8 O2 S
Certifications/Compliance ISO 9001
Synonym 2-Thiophenecarboxylic acid ethyl ester, 2-(Ethoxycarbonyl)thiophene, 2-Thienylcarboxylic acid ethyl ester, Ethyl 2-thenoate, Ethyl 2-thiophenecarboxylate, Ethyl thiophen-2-carboxylate, NSC 39455
InChI Formula InChI=1S/C7H8O2S/c1-2-9-7(8)6-4-3-5-10-6/h3-5H,2H2,1H3
SMILES CCOC(=O)c1cccs1
IUPAC Name ethyl thiophene-2-carboxylate
Molecular Weight (g/mol) 156.2
Quantity 100 mg
Formula Weight 156.0245 g/mol