Läs mer
Ethyl trans-3-Methyl-4-oxocrotonate, TRC
Produktkod. 30369495
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30369495 1 g
1 options
Produktkod. 30369495 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9219851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl trans-3-Methyl-4-oxocrotonate
CAS 62054-49-3
Molekylformel C7H10O3
Kvantitet 1 g
Synonym (E)-3-Methyl-4-oxo-2-butenoic Acid Ethyl Ester, 3-Methyl-fumaraldehydic Acid Ethyl Ester, Ethyl (2E)-3-Methyl-4-oxobut-2-enoate
InChI formel InChI=1S/C7H10O3/c1-3-10-7(9)4-6(2)5-8/h4-5H,3H2,1-2H3/b6-4+
LEDER CCOC(=O)\C=C(/C)\C=O
IUPAC-namn ethyl (E)-3-methyl-4-oxobut-2-enoate
Molekylvikt (g/mol) 142.15
Formel vikt 142.063