missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl trifluoromethanesulfonate, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Ethyl trifluoromethanesulfonate is a powerful ethylating agent.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Ethyl trifluoromethanesulfonate is a powerful ethylating agent.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Specifications
Boiling Point | 115°C to 118°C |
Flash Point | 35°C (95°F) |
Odor | Pungent |
Linear Formula | CF3SO3CH2CH3 |
Quantity | 1 g |
UN Number | UN2920 |
Beilstein | 1770746 |
Formula Weight | 178.13 |
Percent Purity | 99% |
Chemical Name or Material | Ethyl trifluoromethanesulfonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'