Läs mer
Ethyl (Z)-[2-amino-4-thiazolyl](methoxyimino)acetate, TRC
Produktkod. 30378775
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30378775 1 g
1 options
Produktkod. 30378775 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE6784751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl (Z)-[2-amino-4-thiazolyl](methoxyimino)acetate
CAS 64485-88-7
Molekylformel C8H11N3O3S
Kvantitet 1 g
Synonym 2-(2-Amino-4-thiazolyl)-2(Z)(methoxy)imino ethyl acetate, Ethyl (Z)-(2-Aminothiazol-4-yl)-α-methoxyiminoacetate, Ethyl (Z)-[2-Amino-4-thiazolyl](methoxyimino)acetate, Ethyl 2-(2-Aminothiazol-4-yl)-(Z)-2-(methoxyimino)acetate
InChI formel InChI=1S/C8H11N3O3S/c1-3-14-7(12)6(11-13-2)5-4-15-8(9)10-5/h4H,3H2,1-2H3,(H2,9,10)/b11-6-
LEDER O=C(OCC)/C(C1=CSC(N)=N1)=N\OC
Molekylvikt (g/mol) 229.26