Läs mer
Ethylcarbamic Acid 4-Nitrophenyl Ester, TRC
Produktkod. 30372010
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30372010 25 g
1 options
Produktkod. 30372010 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE90088025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethylcarbamic Acid 4-Nitrophenyl Ester
CAS 17576-41-9
Molekylformel C9 H10 N2 O4
Kvantitet 25 g
Synonym Ethylcarbamic Acid p-Nitrophenyl Ester
InChI formel InChI=1S/C9H10N2O4/c1-2-10-9(12)15-8-5-3-7(4-6-8)11(13)14/h3-6H,2H2,1H3,(H,10,12)
LEDER CCNC(=O)Oc1ccc(cc1)[N+](=O)[O-]
IUPAC-namn (4-nitrophenyl) N-ethylcarbamate
Molekylvikt (g/mol) 210.187
Formel vikt 210.064