missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-amino-4-methylthiophene-3-carboxylate, 97%, Thermo Scientific™

Catalog Number p-2704082
Click to view available options
Quantity:
1 g
25 g
This item is not returnable. View return policy
43088-42-2
C8H11NO2S
185.241
MFCD00051669
ILYCZKOBLRJJSW-UHFFFAOYSA-N
2-amino-4-methylthiophene-3-carboxylic acid ethyl ester, 2-amino-4-methyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4-methyl-, ethyl ester, ethyl2-amino-4-methylthiophene-3-carboxylate, ethyl 2-amino-4-methyl-3-thiophenecarboxylate, acmc-20a9ux, maybridge1_003755, 2-amino-4-methyl-3-ethoxycarbonyl-thiophene, ethyl 2-amino4-methylthiophene-3-carboxylate, 2-amino-3-ethoxycarbonyl-4-methylthiophene
521132
ethyl 2-amino-4-methylthiophene-3-carboxylate
CCOC(=O)C1=C(SC=C1C)N
This item is not returnable. View return policy
43088-42-2
C8H11NO2S
185.241
MFCD00051669
ILYCZKOBLRJJSW-UHFFFAOYSA-N
2-amino-4-methylthiophene-3-carboxylic acid ethyl ester, 2-amino-4-methyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4-methyl-, ethyl ester, ethyl2-amino-4-methylthiophene-3-carboxylate, ethyl 2-amino-4-methyl-3-thiophenecarboxylate, acmc-20a9ux, maybridge1_003755, 2-amino-4-methyl-3-ethoxycarbonyl-thiophene, ethyl 2-amino4-methylthiophene-3-carboxylate, 2-amino-3-ethoxycarbonyl-4-methylthiophene
521132
ethyl 2-amino-4-methylthiophene-3-carboxylate
CCOC(=O)C1=C(SC=C1C)N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 185.25
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-methylthiophene-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'