missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromobenzoate, 98%, Thermo Scientific Chemicals

Catalog Number p-3757836
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6091-64-1
C9H9BrO2
229.07
MFCD00015443
BIHHBTVQFPVSTE-UHFFFAOYSA-N
2-bromobenzoic acid ethyl ester, benzoic acid, 2-bromo-, ethyl ester, ethyl-2-bromobenzoate, ethyl2-bromobenzoate, ethyl bromobenzoate, ethyl o-bromobenzoate, pubchem3937, brombenzoesaureathylester, acmc-209mn8, ksc493o4r
80186
ethyl 2-bromobenzoate
CCOC(=O)C1=CC=CC=C1Br
This item is not returnable. View return policy
6091-64-1
C9H9BrO2
229.07
MFCD00015443
BIHHBTVQFPVSTE-UHFFFAOYSA-N
2-bromobenzoic acid ethyl ester, benzoic acid, 2-bromo-, ethyl ester, ethyl-2-bromobenzoate, ethyl2-bromobenzoate, ethyl bromobenzoate, ethyl o-bromobenzoate, pubchem3937, brombenzoesaureathylester, acmc-209mn8, ksc493o4r
80186
ethyl 2-bromobenzoate
CCOC(=O)C1=CC=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.4400g/mL
Boiling Point 117°C (17.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula BrC6H4CO2C2H5
Refractive Index 1.5430 to 1.545
Quantity 25 g
Specific Gravity 1.44
Formula Weight 229.07
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 2-bromobenzoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'