missing translation for 'onlineSavingsMsg'
Learn More

ethyle4-methyl-2-phenyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

Catalog Number p-2704790
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
53715-64-3
C13H13NO2S
247.312
MFCD00141953
FYPLITQTMHJFKK-UHFFFAOYSA-N
ethyl 4-methyl-2-phenylthiazole-5-carboxylate, maybridge1_008302, ethyl4-methyl-2-phenyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-phenyl-5-thiazolecarboxylate, ethyl 4-methyl-2-phenyl thiazole-5-carboxylate, ethyl 4-methyl-2-phenyl-thiazole-5-carboxylate, 5-ethoxycarbonyl-4-methyl-2-phenyl-1,3-thiazole, 2-phenyl-4-methylthiazole-5-carboxylic acid ethyl ester, 4-methyl-2-phenyl-5-thiazolecarboxylic acid ethyl ester, 4-methyl-2-phenyl-thiazole-5-carboxylic acid ethyl ester
2743970
ethyl 4-methyl-2-phenyl-1,3-thiazole-5-carboxylate
CCOC(=O)C1=C(N=C(S1)C2=CC=CC=C2)C
This item is not returnable. View return policy
53715-64-3
C13H13NO2S
247.312
MFCD00141953
FYPLITQTMHJFKK-UHFFFAOYSA-N
ethyl 4-methyl-2-phenylthiazole-5-carboxylate, maybridge1_008302, ethyl4-methyl-2-phenyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-phenyl-5-thiazolecarboxylate, ethyl 4-methyl-2-phenyl thiazole-5-carboxylate, ethyl 4-methyl-2-phenyl-thiazole-5-carboxylate, 5-ethoxycarbonyl-4-methyl-2-phenyl-1,3-thiazole, 2-phenyl-4-methylthiazole-5-carboxylic acid ethyl ester, 4-methyl-2-phenyl-5-thiazolecarboxylic acid ethyl ester, 4-methyl-2-phenyl-thiazole-5-carboxylic acid ethyl ester
2743970
ethyl 4-methyl-2-phenyl-1,3-thiazole-5-carboxylate
CCOC(=O)C1=C(N=C(S1)C2=CC=CC=C2)C
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 247.32
Percent Purity 97%
Chemical Name or Material Ethyl 4-methyl-2-phenyl-1, 3-thiazole-5-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'