missing translation for 'onlineSavingsMsg'
Learn More

ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate, 97%, Thermo Scientific™

Catalog Number p-2704806
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
423768-50-7
C10H8BrNO3S
302.142
MFCD03407336
OIZZMDOBYXRSNQ-UHFFFAOYSA-N
ethyl 5-5-bromo-2-thienyl-3-isoxazolecarboxylate, ethyl 5-5-bromothiophen-2-yl-1,2-oxazole-3-carboxylate, ethyl 5-5-bromothiophen-2-yl isoxazole-3-carboxylate
2776548
ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate
CCOC(=O)C1=NOC(=C1)C2=CC=C(S2)Br
This item is not returnable. View return policy
423768-50-7
C10H8BrNO3S
302.142
MFCD03407336
OIZZMDOBYXRSNQ-UHFFFAOYSA-N
ethyl 5-5-bromo-2-thienyl-3-isoxazolecarboxylate, ethyl 5-5-bromothiophen-2-yl-1,2-oxazole-3-carboxylate, ethyl 5-5-bromothiophen-2-yl isoxazole-3-carboxylate
2776548
ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate
CCOC(=O)C1=NOC(=C1)C2=CC=C(S2)Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 302.15
Percent Purity 97%
Chemical Name or Material ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'