Läs mer
Ethylene bis-Palmitamide, TRC
Produktkod. 30377733
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30377733 100 mg
1 options
Produktkod. 30377733 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE451825100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethylene bis-Palmitamide
CAS 5518-18-3
Molekylformel C34H68N2O2
Kvantitet 100 mg
Synonym N,N'-(Ethane-1,2-diyl)dipalmitamide, 1,2-Bis(hexadecanamido)ethane, N,N'-Dipalmitoylethylenediamine
InChI formel InChI=1S/C34H68N2O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33(37)35-31-32-36-34(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-32H2,1-2H3,(H,35,37)(H,36,38)
LEDER CCCCCCCCCCCCCCCC(=O)NCCNC(=O)CCCCCCCCCCCCCCC
Molekylvikt (g/mol) 536.92
Formel vikt 536.5281