Läs mer
Ethylene Glycol Monopalmitate, TRC
Produktkod. 17929554
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
17929554 50 mg
1 options
Produktkod. 17929554 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE91757550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethylene Glycol Monopalmitate
CAS 4219-49-2
Molekylformel C18 H36 O3
Kvantitet 50 mg
Synonym Ethylene Glycol Monohexadecanoate, 2-Hydroxyethyl Hexadecanoate, 2-Hydroxyethyl Palmitate, Palmitoylglycol
InChI formel InChI=1S/C18H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(20)21-17-16-19/h19H,2-17H2,1H3
LEDER CCCCCCCCCCCCCCCC(=O)OCCO
IUPAC-namn 2-hydroxyethyl hexadecanoate
Molekylvikt (g/mol) 300.48
Formel vikt 300.2664