Läs mer
Ethylenediamine-N,N'-diacetic Acid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Ethylenediamine-N,N'-diacetic acid |
| CAS | 5657-17-0 |
| Molekylformel | C6 H12 N2 O4 |
| Kvantitet | 2.5 g |
| Synonym | Glycine, N,N'-ethylenedi- (6CI,7CI,8CI), N,N'-1,2-Ethanediylbis[glycine], 1,2-Bis(carboxymethylamino)ethane, 1,2-Ethylenediamine-N,N'-diacetic acid, Acetic acid, 2,2'-(1,2-ethanediyldiimino)bis-, EDDA, Ethylenediamine-N,N'-diacetic acid, Ethylenediaminediacetic acid, Ethylenediimino-N,N'-diacetic acid, Ethylenediiminodiacetic acid, N,N'-Bis(carboxymethyl)ethylenediamine, N,N'-Ethylenediaminediacetic acid, N,N'-Ethylenediglycine |
| InChI formel | InChI=1S/C6H12N2O4/c9-5(10)3-7-1-2-8-4-6(11)12/h7-8H,1-4H2,(H,9,10)(H,11,12) |
| LEDER | OC(=O)CNCCNCC(=O)O |
| IUPAC-namn | 2-[2-(carboxymethylamino)ethylamino]acetic acid |
| Molekylvikt (g/mol) | 176.17 |
| Formel vikt | 176.0797 |