missing translation for 'onlineSavingsMsg'
Läs mer

Ethylenediamine-N,N,N’,N’-tetraacetic Acid Calcium Disodium Hydrate, TRC

Produktkod. 30375067
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30375067 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30375067 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE91759010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Edetate Calcium Disodium
CAS 23411-34-9
Molekylformel C10 H12 Ca N2 O8 . 2 Na . H2 O
Kvantitet 10 g
Synonym [(Ethylenedinitrilo)tetraacetato]calciate(2-)Disodium Hydrate
InChI formel InChI=1S/C10H16N2O8.Ca.2Na.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;1H2/q;+2;2*+1;/p-4
LEDER O.[Na+].[Na+].O=C1CN23CCN45CC(=O)[O-][Ca+2]24([O-]C(=O)C5)([O-]C(=O)C3)[O-]1
Molekylvikt (g/mol) 392.28
Formel vikt 392.0121
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.