Läs mer
Ethylmorphine EP Impurity D, Mikromol™
Produktkod. 30050916
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050916 25 mg
30050047 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050916 Leverantör Mikromol Leverantörsnummer MM0799.060025
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethylmorphine EP Impurity D
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethylmorphinone
CAS 93290-69-8
Analyt- eller komponentnamn Ethylmorphinone
Frakt skick Room Temperature
Molekylformel C19 H21 N O3
Certifikat och efterlevnad ISO 17025
Synonym 7,8-Didehydro-4,5α-epoxy-3-ethoxy-17-methylmorphinan-6-one (Ethylmorphinone), (5alpha)-7,8-Didehydro-4,5-epoxy-3-ethoxy-17-methylmorphinan-6-one (9Cl), Ethylmorphinone, Ethylmorphine Hydrochloride Impurity D, Ethylmorphine Impurity D
InChI formel InChI=1S/C19H21NO3/c1-3-22-15-7-4-11-10-13-12-5-6-14(21)18-19(12,8-9-20(13)2)16(11)17(15)23-18/h4-7,12-13,18H,3,8-10H2,1-2H3/t12-,13+,18-,19-/m0/s1
LEDER CCOc1ccc2C[C@@H]3[C@@H]4C=CC(=O)[C@@H]5Oc1c2[C@]45CCN3C
IUPAC-namn (4R,4aR,7aR,12bS)-9-ethoxy-3-methyl-1,2,4,4a,7a,13-hexahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one
Molekylvikt (g/mol) 311.37
Kvantitet 25 mg
Formel vikt 311.1521 g/mol