missing translation for 'onlineSavingsMsg'
Learn More

(Ethylthio)trimethylsilane, 90%, Thermo Scientific Chemicals

Catalog Number p-132363
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5573-62-6
C5H14SSi
134.31
MFCD00042890
HXAFQWICVBBXMZ-UHFFFAOYSA-N
ethylthio trimethylsilane, ethylthiotrimethylsilane, ethylthio trimethylsilane, technical grade, acmc-20aplk, trimethyl ethylthio silane, ethyl thio trimethylsilane, ethyl trimethylsilyl sulfide, trimethylsilyl ethyl sulfide, ethylsulfanyl trimethylsilane, ethylsulfanyl trimethyl silane
2733426
ethylsulfanyl(trimethyl)silane
CCS[Si](C)(C)C
This item is not returnable. View return policy
5573-62-6
C5H14SSi
134.31
MFCD00042890
HXAFQWICVBBXMZ-UHFFFAOYSA-N
ethylthio trimethylsilane, ethylthiotrimethylsilane, ethylthio trimethylsilane, technical grade, acmc-20aplk, trimethyl ethylthio silane, ethyl thio trimethylsilane, ethyl trimethylsilyl sulfide, trimethylsilyl ethyl sulfide, ethylsulfanyl trimethylsilane, ethylsulfanyl trimethyl silane
2733426
ethylsulfanyl(trimethyl)silane
CCS[Si](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Color Colorless
Density 0.8300g/mL
Boiling Point 69°C to 70°C (100.0 mmHg)
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula C2H5SSi(CH3)3
Refractive Index 1.4470 to 1.451
Quantity 1 g
Specific Gravity 0.83
Formula Weight 134.3
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material (Ethylthio)trimethylsilane, 90%
Show More Show Less
compliance-icons
Product Identifier
  • (Ethylthio)trimethylsilane, tech.
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'