Läs mer
Ethynodiol Diacetate, Mikromol™
Produktkod. 30049082
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30049082 250 mg
1 options
Produktkod. 30049082 Leverantör Mikromol Leverantörsnummer MM1960.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethynodiol Diacetate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethynodiol Diacetate
CAS 297-76-7
Analyt- eller komponentnamn Ethynodiol Diacetate
Frakt skick Room Temperature
Molekylformel C24 H32 O4
Certifikat och efterlevnad ISO 17034
Synonym 8080CB, Cervicundin, Continuin, Ethinodiol diacetate, Ethynodiol acetate, Ethynodiol diacetate, Femulen, Luteonorm, Luto-Metrodiol, Metrodiol, Metrodiol, diacetate, SC 11800, 19-Norpregn-4-en-20-yne-3,17-diol, 3,17-diacetate, (3beta,17alpha)-, 19-Nor-17alpha-pregn-4-en-20-yne-3beta,17-diol, diacetate (7CI,8CI), 19-Norpregn-4-en-20-yne-3,17-diol, diacetate, (3beta,17alpha)- (9CI), 17-Ethynyl-19-norandrost-4-ene-3beta,17beta-diol diacetate, 17alpha-Ethynyl-4-estren-3beta,17-diol diacetate, 17alpha-Ethynyl-4-estrene-3beta,17beta-diacetate, 17alpha-Ethynyl-4-estrene-3beta,17beta-diol diacetate, 3beta,17beta-Diacetoxy-19-nor-17alpha-pregn-4-en-20-yne, beta-Ethynodiol diacetate
InChI formel InChI=1S/C24H32O4/c1-5-24(28-16(3)26)13-11-22-21-8-6-17-14-18(27-15(2)25)7-9-19(17)20(21)10-12-23(22,24)4/h1,14,18-22H,6-13H2,2-4H3/t18-,19-,20+,21+,22-,23-,24-/m0/s1
LEDER CC(=O)O[C@H]1CC[C@@H]2[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]4(OC(=O)C)C#C)[C@@H]3CCC2=C1
IUPAC-namn [(3S,8R,9S,10R,13S,14S,17R)-17-acetyloxy-17-ethynyl-13-methyl-2,3,6,7,8,9,10,11,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Molekylvikt (g/mol) 384.51
Kvantitet 250 mg
Formel vikt 384.2301 g/mol