Läs mer
Ethynodiol, TRC
Produktkod. 30386058
Change view
Kvantitet:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30386058 250 mg
30377129 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30386058 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE932120250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1231-93-2
Molekylformel C20 H28 O2
Molekylvikt (g/mol) 300.44
Synonym (3β,17α)-19-Norpregn-4-en-20-yne-3,17-diol, 19-Nor-17α-pregn-4-en-20-yne-3β,17-diol (6CI,7CI,8CI), 17α-Ethynyl-4-estrene-3β,17β-diol, 17α-Ethynyl-4-oestrene-3β,17β-diol, 3β-Hydroxylynestrenol, Ethinodiol, Ethynodiol, Etynodiol
IUPAC-namn (3S,8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-2,3,6,7,8,9,10,11,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthrene-3,17-diol
LEDER C[C@]12CC[C@H]3[C@@H](CCC4=C[C@@H](O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
Specifikationer
Kemiskt namn eller material Ethynodiol
Kvantitet 250 mg
InChI formel InChI=1S/C20H28O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,12,14-18,21-22H,4-11H2,2H3/t14-,15-,16+,17+,18-,19-,20-/m0/s1
Formel vikt 300.2089