Läs mer
Ethynyl-2-13C-benzene, TRC
Produktkod. 30134166
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134166 100 mg
30007215 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134166 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE932154100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54522-92-8
Molekylformel 13C C7 H6
Molekylvikt (g/mol) 103.13
Synonym Benzene, ethynyl-2-13C- (6CI, 9CI, ACI), Ethynyl-2-13Cbenzene (ACI), Ethynyl-2-13C-benzene, Ethynylbenzene-13C (ethynyl-2-13C), Phenylacetylene-2-13C, Phenylacetylene-13C
IUPAC-namn (2^{13}C)ethynylbenzene
LEDER [13CH]#Cc1ccccc1
Specifikationer
Kemiskt namn eller material Ethynyl-2-13C-benzene
Kvantitet 100 mg
InChI formel InChI=1S/C8H6/c1-2-8-6-4-3-5-7-8/h1,3-7H/i1+1
Formel vikt 103.0503