Läs mer
Ethynyl Estradiol 17-Acetate, TRC
Produktkod. 30379864
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30379864 10 mg
30376344 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30379864 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE68513010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethynyl Estradiol 17-Acetate
CAS 21221-29-4
Molekylformel C22 H26 O3
Kvantitet 10 mg
Synonym 19-Norpregna-1,3,5(10)-trien-20-yne-3,17-diol, 17-acetate, (17α)-, 19-Nor-17α-pregna-1,3,5(10)-trien-20-yne-3,17-diol, 17-acetate (8CI), 17-Ethynylestra-1,3,5(10)-triene-3,17β-diol 17-acetate
InChI formel InChI=1S/C22H26O3/c1-4-22(25-14(2)23)12-10-20-19-7-5-15-13-16(24)6-8-17(15)18(19)9-11-21(20,22)3/h1,6,8,13,18-20,24H,5,7,9-12H2,2-3H3/t18-,19-,20+,21+,22+/m1/s1
LEDER CC(=O)O[C@]1(CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C)C#C
IUPAC-namn [(8R,9S,13S,14S,17R)-17-ethynyl-3-hydroxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl] acetate
Molekylvikt (g/mol) 338.44
Formel vikt 338.1882