Läs mer
Etilefrine Hydrochloride, LoGiCal
Produktkod. 30461720
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30461720 10 mg
1 options
Produktkod. 30461720 Leverantör LoGiCal Leverantörsnummer LGCFOR0035.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Etilefrine hydrochloride
CAS 943-17-9
Analyt- eller komponentnamn Etilefrine hydrochloride
Frakt skick Room Temperature
Molekylformel C10 H15 N O2 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym 1-(3-Hydroxyphenyl)-2-ethylaminoethanol hydrochloride, Apocretin, Circupon, Effontil, Effortil, Effortilvet, Ethyl adrianol, Ethylephrine hydrochloride, Eti-Puren, Etilefrine hydrochloride, Funasol, Kertasin, Phetanol, Pulsamin, Tonus-Forte, Updormin, dl-1-(3-Hydroxyphenyl)-1-hydroxy-2-ethylaminoethane hydrochloride, dl-1-(3-Hydroxyphenyl)-2-ethylaminoethanol hydrochloride, dl-Effortil hydrochloride, dl-Ethylphenylephrine hydrochloride, dl-Etilefrin hydrochloride, dl-N-Ethylnorphenylephrine hydrochloride, Benzenemethanol, alpha-[(ethylamino)methyl]-3-hydroxy-, hydrochloride (1:1), Benzenemethanol, alpha-[(ethylamino)methyl]-3-hydroxy-, hydrochloride (9CI), Benzyl alcohol, alpha-[(ethylamino)methyl]-m-hydroxy-, hydrochloride (6CI,8CI), (+/-)-Ethylphenylephrine hydrochloride, (+/-)-N-Ethylnorphenylephrine hydrochloride, alpha-[(Ethylamino)methyl]-m-hydroxybenzyl alcohol hydrochloride
InChI formel InChI=1S/C10H15NO2.ClH/c1-2-11-7-10(13)8-4-3-5-9(12)6-8;/h3-6,10-13H,2,7H2,1H3;1H
Lösningstyp Neat
LEDER Cl.CCNCC(O)c1cccc(O)c1
IUPAC-namn 3-[2-(ethylamino)-1-hydroxyethyl]phenol;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 217.69
Kvantitet 10 mg
Formel vikt 217.087 g/mol