Läs mer
Etilefrine Hydrochloride, Mikromol™
Beskrivning
Etilefrine Hydrochloride
Specifikationer
Specifikationer
| Kemiskt namn eller material | Etilefrine hydrochloride |
| CAS | 943-17-9 |
| Analyt- eller komponentnamn | Etilefrine hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C10 H15 N O2 . Cl H |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | 1-(3-Hydroxyphenyl)-2-ethylaminoethanol hydrochloride, Apocretin, Circupon, Effontil, Effortil, Effortilvet, Ethyl adrianol, Ethylephrine hydrochloride, Eti-Puren, Etilefrine hydrochloride, Funasol, Kertasin, Phetanol, Pulsamin, Tonus-Forte, Updormin, dl-1-(3-Hydroxyphenyl)-1-hydroxy-2-ethylaminoethane hydrochloride, dl-1-(3-Hydroxyphenyl)-2-ethylaminoethanol hydrochloride, dl-Effortil hydrochloride, dl-Ethylphenylephrine hydrochloride, dl-Etilefrin hydrochloride, dl-N-Ethylnorphenylephrine hydrochloride, Benzenemethanol, alpha-[(ethylamino)methyl]-3-hydroxy-, hydrochloride (1:1), Benzenemethanol, alpha-[(ethylamino)methyl]-3-hydroxy-, hydrochloride (9CI), Benzyl alcohol, alpha-[(ethylamino)methyl]-m-hydroxy-, hydrochloride (6CI,8CI), (+/-)-Ethylphenylephrine hydrochloride, (+/-)-N-Ethylnorphenylephrine hydrochloride, alpha-[(Ethylamino)methyl]-m-hydroxybenzyl alcohol hydrochloride |
| InChI formel | InChI=1S/C10H15NO2.ClH/c1-2-11-7-10(13)8-4-3-5-9(12)6-8;/h3-6,10-13H,2,7H2,1H3;1H |
| LEDER | Cl.CCNCC(O)c1cccc(O)c1 |
| IUPAC-namn | 3-[2-(ethylamino)-1-hydroxyethyl]phenol;hydrochloride |
| Visa mer |