Läs mer
Etodolac Ethyl Ester, Mikromol™
Produktkod. 30051871
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051871 25 mg
30049344 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051871 Leverantör Mikromol Leverantörsnummer MM0911.130025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Etodolac Ethyl Ester
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Etodolac Ethyl Ester
CAS 200880-23-5
Analyt- eller komponentnamn Etodolac Ethyl Ester
Frakt skick Room Temperature
Molekylformel C19 H25 N O3
Certifikat och efterlevnad ISO 17025
Synonym Etodolac Ethyl Ester, 1,8-Diethyl-1,3,4,9-tetrahydro-pyrano[3,4-b]indole-1-acetic acid ethyl ester
InChI formel InChI=1S/C19H25NO3/c1-4-13-8-7-9-14-15-10-11-23-19(5-2,12-16(21)22-6-3)18(15)20-17(13)14/h7-9,20H,4-6,10-12H2,1-3H3
LEDER CCOC(=O)CC1(CC)OCCc2c1[nH]c3c(CC)cccc23
IUPAC-namn ethyl 2-(1,8-diethyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetate
Molekylvikt (g/mol) 315.41
Kvantitet 25 mg
Formel vikt 315.1834 g/mol