Läs mer
Etodolac, MedChemExpress
Produktkod. 16457946
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16457946 100 mg
16434107 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16457946 Leverantör Medchem Express Leverantörsnummer HY76251
MedChemExpress Etodolac (AY-24236) is a non-steroidal anti-inflammatory compound that is a non-selective inhibitor of COX (IC50=53.5 nM)
Etodolac (AY-24236) is a non-steroidal anti-inflammatory compound that is a non-selective inhibitor of COX (IC50=53.5 nM)
Specifikationer
Kemiskt namn eller material Etodolac
Fysisk form Solid
CAS 41340-25-4
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C17H21NO3
För användning med (applikation) COVID-19-immunoregulation
Synonym AY-24236
Löslighetsinformation DMSO : ≥ 100 mg/mL (348.01 mM) ∣H2O : < 0.1 mg/mL (insoluble)
LEDER O=C(O)CC(OCC1)(CC)C2=C1C3=CC=CC(CC)=C3N2
Molekylvikt (g/mol) 287.35
Formel vikt 287.35
Procent renhet 99.11%